Organic Chemistry Set
Supplier: GSC International
ORGCHEM-19-22
470313-394EA 268.3 USD
470313-394
Organic Chemistry Set
Labware Sets
This set is made from high-quality borosilicate glass and all joints are ground 19/22.
Set Includes: (1) West condenser, (1) Distilling Column, (5) Round-bottom flask (500mm, 250mm, 100mm, 50mm, 25mm), (1) Separatory funnel, 100ml, (1) Bleed tube, (1) Thermometer holder, (1) Thermometer adapter, (1) Three-way adapter, (1) Vacuum adapter, (1) Claisen adapter, (1) Stopper.