G.S.C. International Filtering Flask Vacuum Pump Kit
Supplier: GSC International
4400
470313-372EA 82 USD
470313-372
G.S.C. International Filtering Flask Vacuum Pump Kit
Pumps Vacuum Pumps
More economical than an electric vacuum pump.
This Filtration Kit allows rapid separation of mixtures via vacuum filtration. The stopper is cut to hold a Buchner funnel. The hand-operated vacuum pump simplifies vacuum filtration, eliminating the need for a water source or vacuum aspirator. The pumping rate is 15 ml per stroke, and will displace up to 725 ml of air. The pump is fitted with a valve to release the vacuum.
Delivery information: Kit includes borosilicate filtering flask (500 ml), vacuum pump, Buchner funnel, filter paper (7 cm), connection tubing and rubber stopper.