Alcohol Burner Dual Wick
Supplier: GSC International
42002
470313-338EA 33.4 USD
470313-338
Alcohol Burner Dual Wick
Heaters Spirit Lamps
This all metal, permanent wick alcohol burner has a tank capacity of 100 ml.
The 'wick' portion is a ceramic fiber inserted into a thin copper tube. This construction allows for a constant flame. The burner is fueled by high-quality alcohol which can be found in most local hardware stores (not drug store rubbing alcohol). This is an excellent alternative to the traditional glass alcohol burners because it is durable and less likely to spill. Instructions included.