Deflagration Spoon
Supplier: GSC International
4-32901-12
470313-336EA 10.9 USD
470313-336
Deflagration Spoon
Spoons
Deflagration Spoons are commonly used in experiments that require students to burn phosphorous, sulfur, or other substances at a safe distance.
The handle is angled to keep hands away from the heat source. It is 18" long with a 3/4" diameter bowl and 1/4" depth.