12-Piece Cork Borer Set
Supplier: GSC International
4-CBS-12
470313-326EA 70.5 USD
470313-326
12-Piece Cork Borer Set
Cork Borers
The cork borer set includes 12 borers, in sizes ranging from 4.8 to 21 mm.
  • Borers are made of nickel-plated steel
  • Ergonomic handles provide a good grip on the tool
  • Hole diameters range between 4.8 and 21 mm