Bird and Mammal Bone Comparison
Supplier: American 3B Scientific
1021045
470312-620EA 819.95 USD
470312-620
Bird and Mammal Bone Comparison
Models Zoology Models
Longitudinal and cross section of a prepared, real bird bone and mammal bone respectively, mounted on a base plate. The product illustrates the difference between the long bones of a bird and a mammal.
  • High quality natural bone