1-Hole Natural Rubber Stoppers
Supplier: United Scientific Supplies
RST1-H
470312-548LB 19.9 USD
470312-548
1-Hole Natural Rubber Stoppers
Stoppers Rubber Stoppers
Superior Grade Rubber Offers Softness, Resiliency, and Durability.
  • Size numbers molded onto the top
  • Low sulfur content
  • All stoppers are 25 mm long
Designed to stay pliable over long period of time, these stoppers can withstand temperatures from –25 to 70°C (–13 to 158°F). Ideal for use with nonaggressive or concentrated solutions.
Caution: Item contains latex, which is known to cause allergic reactions in some people. Use with caution.