Somso® Maize Leaf Ls Cs
Supplier: MARCUS SOMMER
BOS17/3
470312-462EA 1090.85 USD
470312-462
Somso® Maize Leaf Ls Cs
Models Botany Models
Somso® premium quality models.
  • 5 year warranty
Zea mays, enlarged approx. 450 times, in SOMSO-Plast®. After Prof. Dr. W. Weber. The model shows the special leaf structure of a C4 plant. The vascular bundles are enclosed by a sheath in ring form. The chloroplasts of the sheath correspond with the biochemical work appropriation in the case of photosynthesis and are clearly larger than the chloroplasts of the mesophyll cells. The vascular bundles are interlinked with each other by transversal anastomoses. In one piece, on a stand with green base and description key.
Ordering information: Items on this page are not stocked and may have an extended time for delivery.