Somso® Real Camomile
Supplier: MARCUS SOMMER
BOS15/6
470312-454EA 503.59 USD
470312-454
Somso® Real Camomile
Models Botany Models
Somso® premium quality models.
  • 5 year warranty
Matricaria chamomilla, inflorescene (composite), enlarged approx. 9 times, in SOMSO-Plast®. Ligulate flower 20x magnification, tubular flower 80x magnification. After Prof. Dr. W. Weber. In one piece. On a stand with green base and explanatory note.
Ordering information: Items on this page are not stocked and may have an extended time for delivery.