Somso® Rye Spikelet
Supplier: MARCUS SOMMER
BOS15/5
470312-452EA 616.39 USD
470312-452
Somso® Rye Spikelet
Models Botany Models
Somso® premium quality models.
  • 5 year warranty
Secale cereale, enlarged approx. 25 times, in SOMSO-Plast®. After Prof. Dr. W. Jung and Prof. Dr. W. Weber. Separable into 4 parts to show the typical structure of an earlet of grass. On a stand with green base and explanatory note.
Ordering information: Items on this page are not stocked and may have an extended time for delivery.