Somso® Primrose
Supplier: MARCUS SOMMER
BOS15/4
470312-450EA 656.59 USD
470312-450
Somso® Primrose
Models Botany Models
Somso® premium quality models.
  • 5 year warranty
Primula officinalis (cowslip), in SOMSO-Plast®. After Prof. Dr. W. Weber. Median section through two heterostyled smelling primrose flowers, enlarged approx. 13 times. The relief presentation highlights the heterostyly to facilitate cross fertilization. In one piece. On a stand and green base with explanatory note.
Ordering information: Items on this page are not stocked and may have an extended time for delivery.