VersaLoupe™ LH-30 Magnifier, Carson
Supplier: CARSON OPTICAL
LH-30
470312-386EA 13.1 USD
470312-386
VersaLoupe™ LH-30 Magnifier, Carson
Magnifiers
Accurate and easy to focus.
The LH-30 VersaLoupe™ is a 10x dual lens stand magnifier. It is a crystal clear acrylic loupe with an adjustable focus. Simply twist the eyepiece to focus. It is perfect for coins, stamps, and photography.