3B Scientific® Lecture Hall Ear
Supplier: American 3B Scientific
1001266
470311-834EA 12841 USD
470311-834
3B Scientific® Lecture Hall Ear
Models Human Biology Models
Is this possibly the worlds largest ear?
  • 15x life size
  • Includes base
  • Three year manufacturers warranty
  • Includes 3B Smart Anatomy
At 15 times life-size, this 3-part human ear is suitable for museums and special collections as well as large lecture halls and conferences. The giant ear represents the outer, middle and inner ear. The auditory ossicles and the labyrinth with cochlea and vestibulocochlear nerve can be removed from the ear and studied in detail.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.