LilyPad MP3
Supplier: SPARKLE LABS
SL031
470311-304EA 80.19 USD
470311-304
LilyPad MP3
Educational Materials Arts and Crafts Kits and Supplies
The LilyPad MP3 player is your all-in-one audio solution, containing an Arduino-compatible microcontroller, MP3 (and many other formats) audio decoder chip, micro-SD card socket, and a stereo audio amplifier. Part of the Lilypad e-textiles line, this board can be used to give your fabric creations the gift of music, or any other sounds you can put on a micro-SD card.
  • Works out of the box
  • Programmable for expert users
It can be powered by a 3.7 V Lithium Potassium battery (charging circuit built-in!), or an external 3.5 to 6 V source. Off the shelf, it will play specific audio files when any of its five trigger inputs are grounded, or you can solder in a RGB rotary encoder (not included) and load new firmware to add a user interface for track selection and volume control!
The board is compatible with sketches written for the MP3 Player Shield. Our example code uses Bill Porter’s MP3 Player library, which makes writing new code very easy. All you need to do is add a microSD card and some speakers, and you are ready to rock.