LilyPad Simple Snap
Supplier: SPARKLE LABS
SL022
470311-302EA 37.69 USD
470311-302
LilyPad Simple Snap
Educational Materials Arts and Crafts Kits and Supplies
The LilyPad SimpleSnap is a new, easy way to create e-textiles projects with LilyPad that are modular and can be disassembled or broken down, it’s also a great way to prototype! The SimpleSnap board is similar to the LilyPad Simple board, and has the same functionality except for two major differences: A built-in rechargeable Lithium Polymer battery and female snap connectors.
  • A built-in rechargeable lithium polymer battery and female snap connectors
  • Easy to prototype then take apart
  • Board can be shared with multiple projects
By adding fabric snaps to the board, LilyPad made it possible to connect this board to the SimpleSnap Protoboard or simply an arrangement of sew-on fabric snaps so that the board is removable from your project for washing or so that multiple projects can share a board! It is important, though, that you do not wash the SimpleSnap because the battery can be damaged.
The on-board Lithium Polymer battery can be charged simply by attaching an FTDI breakout (the same board used for programming) and just like the Simple, the SimpleSnap can be programmed in Arduino!