Rainbow LED Pack
Supplier: SPARKLE LABS
SL010
470311-296PK 11.3 USD
470311-296
Rainbow LED Pack
Educational Materials Arts and Crafts Kits and Supplies
A rainbow of colors in nine clear 5 mm LEDs and one white 10 mm LED. These are standard, super-bright LEDs. You can use them with a 3 V power supply or in series with a resistor and a 5 V power supply.
  • Clear LEDs that come in lime, blue, pink, purple, yellow, aqua, orange, red, green and white
  • Breadboard ready
  • Beautiful colors
Nine colors included plus one bonus 10 mm LED: red, orange, yellow, lime, green, blue, aqua, violet, white. Let us make some rainbows. 5 mm LEDs. Great for breadboards too.