Sewable LED Stars
Supplier: SPARKLE LABS
SL002
470311-286PK 5.3 USD
470311-286
Sewable LED Stars
Educational Materials Arts and Crafts Kits and Supplies
Star shaped white LED light for sewing onto to something. These stars can be used without a microcontroller but require the sewable battery holder. Can be used with the LilyPad or other textile ready microcontroller.
  • Fun design
  • Resistors are on the board
  • Larger size so they can be easily sewn
These cool LEDs have a built-in resistor. They rely on conductive thread. Our high-quality conductive thread and sewable battery holder are perfect companions to these sewable stars. With the battery removed, fabrics with electronics can be washed. Conceptually, these circuits are identical to their wire and solder counterparts, observing the same principles of polarity and resistance. Only the materials, methods and sometimes scale are different. There is a lot more to sewable circuits than LEDs. Check out the Sparkle Labs E-Textile kit or the E-Textile Lab pack.