Indirect Vent Safety Goggles
Supplier: VWR International
470311-272
470311-272PK 104.25 USD
470311-272
Indirect Vent Safety Goggles
Goggles
These economical goggles protect students from chemical splashes and impact with a flexible ridge around a PVC frame.
  • Indirect ventilation
  • Quad vents to minimize lens fogging
  • Goggles fit over most prescription glasses
  • Antifog coating available
They feature hooded, quad, and indirect vents to minimize lens fogging and provide optimal peripheral vision.
Certifications: Meet ANSI Z87+ 1-2010 standards.
Ordering information: Safety goggle classroom kit includes 31 uncoated indirect vent goggles, 30 student goggles, and 1 teacher's pair of goggles (all 470016-082).