The Future of Biofuels - Alcohol Fermentation, Edvotek
Supplier: EDVOTEK
304
470311-248EA 179 USD
470311-248
The Future of Biofuels - Alcohol Fermentation, Edvotek
Educational Classroom Kits and Activities
Produce ethanol biofuel through yeast fermentation of sugar in different environmental conditions.
  • Make biofuel directly in the classroom
  • For five lab groups
  • Complete activity in 5 to 6 hours
Ethanol fermentation is the most common method for biofuel production worldwide. In this experiment, students will use small-scale flask fermenters to quantify ethanol production and sugar utilization by Saccharomyces cerevisiae. By controlling variables such as temperature and aeration, the students can compare the efficiency of the fermentations over a three day experiment.
Ordering information: Stir plate and stir bars, thermometer, graduated cylinders, erlenmeyer flasks, 70% ethanol, distilled water, waterbath (99 °C), air pump (optional), autoclave or oven (optional), centrifuge (optional), spectrophotometer (optional) is required.
Delivery information: Kit includes: instructions, Yeast Microbead, yeast growth media concentrate, glucose concentrate, ampicillin, yeast growth media, 10% glucose solution, Benedict's Reagents, transfer pipets, snap top microcentrifuge tubes, screw top microcentrifuge tubes, pH paper, hydrometer, 50 ml and 15 ml centrifuge tubes.