Thermal Analysis of Hydrocarbons, Leybold Products
Supplier: Klinger
C2.1.2.1
470311-206EA 9255.79 USD
470311-206
Thermal Analysis of Hydrocarbons, Leybold Products
Educational Materials Chemistry Educational Materials Properties of Matter Learning Activities in Chemistry
Leybold Chemistry Experiments.
  • Composition of organic compounds
  • Reactions in organic chemistry
  • Petrochemistry
  • Synthesis and purification of organic compounds
In order to determine the number of hydrogen atoms as compared with the number of carbon atoms, hydrocarbons can be separated into their elements by thermal decomposition. With gaseous hydrocarbons, this can be done with a filament in a combustion chamber. There the gas is split into hydrogen and carbon black (soot). The ratio of hydrogen to carbon in the compound can be derived directly from the increase in gas volume. In experiment C2.1.2.1, the gases methane and ethane are studied in this way.