Quantitative Determination of Hydrogen, Leybold Products
Supplier: Klinger
C2.1.1.2
470311-204EA 9242.99 USD
470311-204
Quantitative Determination of Hydrogen, Leybold Products
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Leybold Chemistry Experiments.
  • Determine the composition of organic compounds
  • Learn the process of thermal analysis
  • Analyze the content of hydrogen in a compound
  • Determine the structure of organic compounds
In experiment C2.1.1.2, the hydrogen content of an organic compound is determined quantitatively. Here, too, the combustion analysis is carried out, whereby the carbon is converted to CO2 and the hydrogen is converted to H2O. The water content is determined from the difference in the weight of the drying tube, and the share of hydrogen is determined from the water content.