Ward's Artic Squares® Freezer Rack
Supplier: VWR International
470311-190
470311-190CS 865 USD
470311-190
Ward's Artic Squares® Freezer Rack
Racks Cryogenic Box Racks
Innovative horizonal drawer system for upright freezers.
  • Constructed of corrosion-resistant stainless steel with four drawers to hold 16 boxes
  • Fits most standard upright freezers
The ward's artic Squares® freezer rack features drawers that have a unique stop, so that when fully extended they pivot down allowing for easy box access. The polycarbonate drawer handles provide a user-friendly grip.
Each rack also includes a slot and label for drawer/sample identification. The freezer track also includes a pull-out wire handle for easy removal off the freezer shelf.