Ward's Cube Upright Freezer Rack
Supplier: VWR International
470311-188
470311-188CS 713.55 USD
470311-188
Ward's Cube Upright Freezer Rack
Racks Cryogenic Box Racks
Designed for 15 and 50 ml tube box storage.
  • Constructed of corrosion-resistant stainless steel with 6 rack spaces
  • Compatible with 6" (15.2 cm) or smaller boxes
The ward's cube upright freezer racks provide a safe and convenient way to store samples. With handles on each end, this rack accommodates freezers with right or left handed doors.