TruBlu™ LED Transilluminator, Edvotek
Supplier: EDVOTEK
557
470311-170EA 460 USD
470311-170
TruBlu™ LED Transilluminator, Edvotek
Transilluminators LED Transilluminators
Includes both blue light and white light functionality all in one unit.
  • Dual Blue/White Light Modes
  • Large Viewing Surface: 27 x 15 cm
  • Orange Contrast Lid
  • Safe - Non-UV Wavelength
  • Built-in Fan Minimizes Condensation
The successor to the popular TruBlu™ has been improved with an enlarged viewing surface and a novel white light mode. Blue light enables visualization of SYBR® Safe stained DNA gels and white light enhances visualization of blue stained DNA or protein gels. The TruBlu™ 2 has enough surface area to simultaneously view up to eight 7 x 7 cm gels and combines the functions of two units into one!