Fractional Distillation Apparatus
Supplier: Wards
CH0890B
470310-688EA 256.85 USD
470310-688
Fractional Distillation Apparatus
Distillation Components Distillation Sets
Fractional distillation process is the separation of a mixture into individual parts of fractions by heating it up.
  • All pieces are made of high quality borosilicate glass 3.3
Delivery information: Kit includes 250 ml Erlenmeyer flask (1), 250 ml round bottom boiling flask (1), Vigreux column (1), Liebig condenser (1), receiver adapter vacuum (1), joint clips (4). Clamps, stand, thermometer or Bunsen burner are not included with this kit.