Simple Distillation Apparatus
Supplier: Wards
CH0886
470310-686EA 75.6 USD
470310-686
Simple Distillation Apparatus
Distillation Components Distillation Sets
This simple distillation apparatus is made up of boroscilicate glass.
Delivery information: Apparatus includes round bottom flask (500 ml), still head with thermometer support, Liebig condensor (300 mm), receiver adapter and 3×joint clip (19/26).