Diamond Saw Lubricant
Supplier: COVINGTON ENGINEERING CORP
R80100
470310-428EA 39.19 USD
470310-428
Diamond Saw Lubricant
Lubricants
This mineral based oil has no distasteful odor and features excellent cooling properties that extend the life of the blade and saw.
  • High flashpoint of 230° F ensures safe operation in all conditions
We recommend this oil for all saws over 10" for cutting hard stone.