Flir One Pro Thermal Imaging Camera Attachment, Flir Commercial Systems INC SE
Supplier: Extech
435-0006-03
470310-242EA 560.49 USD
470310-242 470310-244
Flir One Pro Thermal Imaging Camera Attachment, Flir Commercial Systems INC SE
Cameras
Turn your smartphone into a brilliant phone.
  • Measure temperature accurately and from a safe distance [-4 °C and 752 °F (-20 to 400 °C)]
  • MSX and VividIR enhanced image processing and video capabilities give you the sharpest mobile thermal imaging performance
FREE FLIR ONE mobile app (recently updated) with exclusive content, real-time professional tips and tricks, multiple spot meters, and easy automated reporting to document your findings - USER REGISTRATION NOT REQUIRED.
The FLIR ONE Pro helps you find invisible problems faster than ever, whether you're inspecting electrical panels, troubleshooting mechanical systems, looking for HVAC problems, or finding water damage. Measure temperatures up to 400 °C (752 °F) - with a sensitivity that detects temperature differences down to 70 mK. Packed with powerful measurement tools, the FLIR ONE Pro will work as hard as you do.