USB Humidity/Temperature Datalogger
Supplier: NEW GENIE GROUP, INC MS
RHT30
470310-226EA 211.19 USD
470310-226
USB Humidity/Temperature Datalogger
Data Loggers
Easy-to-use datalogger records humidity and temperature with date/time stamp.
  • User programmable settings: six languages, sample rate, start delay time, alarm delay time, high/low alarm range, and security feature
  • Start button begins continuous datalogging, which continues until memory is full at 48000 readings (24000 for each parameter: humidity/temperature), or when stop button is pressed
  • Bookmark eight locations in a data recording log with a press of the mark button
  • Generates a PDF report or Excel® spreadsheet with data and trending graph when plugged into a PC after datalogging is completed (Adobe® Reader® required)
The datalogger is designed with a rugged housing and a standard USB connector where stored readings (up to 48,000 data) are conveniently transferred to a PC in PDF or Excel® spreadsheet format when plugged in. Additional features include an 8-location logger bookmark, 6-language options, programmable sampling interval, adjustable alarm, and password-protected security log-in. Large LCD optimizes viewing and the smart LED provides color-coded humidity, and temperature reading status.
Delivery information: Supplied with 2 CR2032 batteries, USB cover (can be used as a stand), wall mounting screw, and built-in hanger.