LaCrosse Glass Rain Gauge
Supplier: LA CROSSE TECHNOLOGY
704-1514
470309-888EA 6.5 USD
470309-888
LaCrosse Glass Rain Gauge
Weather Stations
Clear glass rain gauge measures 5" (12cm) of rainfall and can be mounted either onto a post or placed into the ground. Large inch digits and clear measuring lines make it easy to read and removable glass tube for easy cleaning.
  • Measures rainfall in inches/centimeters
  • Mount can be secured to post or staked into ground
  • Aligned with GLOBE protocols