Digital Differential Counter
Supplier: LW Scientific
CTL-DIFD-08KP
470309-548EA 323.2 USD
470309-548
Digital Differential Counter
Cell Counters and Flow Cytometers Electronic Cell Counters
Automatic calculation of percentages.
  • 3 Digit LCD and total count windows
  • Audible click to confirm count record
  • Beep alarm when 100 cell count is reached
  • Unit dimensions: 10L×3.75W×1.75"H
The Digital Differential Counter uses modern technology and microprocessor-controlled electronics for accurate cell counts. The 8 alternating red and white buttons have a comfortable, ergonomic feel for positive and accurate finger positioning, and the 3-digit digital windows display each cell count.
The 8 buttons are labeled with cell names and cell pictures for BASO, EOSIN, MYELO, JUVEN, STAB, SEG, LYMPH, and MONO. A click sound on each touch confirms the count, and a short beep indicates users have reached 100 cells. Push the percent button at any time to automatically calculate percentages, and then keep counting as high as 999 total cells. Mistakes are easy to correct. Simply push the +/- button to subtract the count, and then push the +/- button again to resume counting. Use the included power cord when in the lab, or take the Digital Differential Counter into the field by installing 2 AA batteries. Push the function (FUN) button to adjust the contrast and to monitor the battery energy. The new Digital Differential Counter will improve accuracy, efficiency, and comfort when counting cells in the laboratory.