Makey Makey Classic EDU
Supplier: THE DOUGLAS STEWART CO.,INC
122800
470309-308EA 53.99 USD
470309-308
Makey Makey Classic EDU
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Turn everyday objects like bananas into touchpads.
  • Using the Makey Makey you can make anything into a key just by connecting a few alligator clips
  • It can also act like a keyboard or mouse
  • There are six inputs on the front of the board, which can be attached to via alligator clipping, soldering to the pads, or any other method
Connect the world around you to your computer. Setup takes just seconds. Just plug in, clip, and play. No programming knowledge needed, and no software to install. Works with Mac and Windows.
Ordering information: Includes Makey Makey Board, USB cable, 7 alligator clips, 6 connector wires, instructions with visual project start guide, online documentation and 20 color stickers.