Water Sampler
Supplier: Science First
15010
470308-880EA 103 USD
470308-880
Water Sampler
Samplers Water Samplers
Water sampler for samples of lake, river, or stream water.
  • Collect water samples for quality testing
Efficient, economical water sampler. A single line deploys and triggers the sampler from any depth in lakes, rivers, or ponds. The flow-through design allows a valid sampling for measuring temperature and water quality.
Delivery information: A 14 m line is included.
Caution: Volume may vary slightly. It is recommended that users verify the exact volume of the bottle prior to use.