Dual Range QAC Test Strips, LaMotte®
Supplier: LA MOTTE CHEMICAL PROD CO SE
2934
470308-466EA 11.5 USD
470308-466
Dual Range QAC Test Strips, LaMotte®
Environmental Test Kits
The New Dual Range QAC Test Strip takes care of ALL of your testing needs on ONE STRIP!
  • Low Range: 0, 10, 20, 40, 80
  • High Range: 0, 100, 200, 400, 800
Whether checking the concentration of your QAC solution or checking to ensure complete removal, the new Dual Range QAC Test Strip DOES IT ALL!
Chemically-treated test strips change color to indicate sanitizer level. Strips and color chart are packaged in a waterproof plastic vial.