Ward's® Kick Net
Supplier: Shiv Dial Sud & Sons
B11235H/4A B11235N/9 B11235C/11
470308-194CS 717.56 USD
470308-194 470308-226 470308-238
Ward's® Kick Net
Animal Supplies Animal Nets
For drag style collecting.
  • Durable netting
  • Easily stored and dried
  • Simple assembly in field
  • Mesh size is 200 microns
  • Aligned with GLOBE protocols
The Ward's Kick Net is everything you could want in a student kick net. The square meter collecting area is large enough to collect sufficient samples, but small enough for a single student to manage if necessary, although two students are best. Each end is stabilized by a large pine pole that won’t collapse in a current. The base and sides of the net are framed in muslin sleeves, giving god support and net control.