Ward's® Silicone Lab Mat
Supplier: Heathrow Scientific
120599W
470306-958CS 506.45 USD
470306-958
Ward's® Silicone Lab Mat
Surface Protectors Bench Protectors
Mat features magnetic area to keep stir bars in place. It keeps benchtop clean and safe from stains, spills and wear.
  • Reusable
  • Easy to clean and store
Anti-skid working surface keeps items in place and reduces sound and vibration from equipment.