Hill-Rom P1900 TotalCare Hospital Bed
Supplier: GUMBO MEDICAL
HRT1900
470306-920EA 3865.29 USD
470306-920
Hill-Rom P1900 TotalCare Hospital Bed
Gurneys
Made for acute patient care.
  • Fully refurbished
  • Multi-position bed
  • Fully functioning standard features
  • Includes mattress
The Hill-Rom® TotalCare® Model P1900 bed provides maximum comfort to patients in critical condition and greatly reduces stain to caregivers. This bed features both hydraulic and manual controls which the caregiver can easily transition the patient surface into numerous positions from either side using Point-Of-Care™ siderail controls and HandsFree™ foot controls. The hydraulic movements provide for smoother bed positioning for patients.
The bed comes standard with a Short Stay Surface to provide patient pressure reduction or optional Treatment Air Surface that provides continuous full-body pressure relief management through automatic adjustments in air system to accommodate changes in weight distribution. Furthermore, TotalCare® offers many optional full interaction caregiver controls through a Graphical Control Interface (GCI) which operate optional functions such as bed exit alarm, integrated scale functions, foot elevation and air mattress system.
In addition, this bed offers three chair positions (chair, chair egress and recliner) and FlexAfoot™ retractable foot mechanism to extend or retract foot section. Also, battery backup is available when there is a power loss to temporarily operate the bed positioning functions. Due to its versatility, numerous features, and ease of use in emergency situations, this bed is ideal for acute patient care, medical/surgical care, post anesthesia care, and Intensive Care Units.
Ordering information: Additional features and models may be available. Please contact your sales representative or customer service for information. This is a large item and will require freight shipment. Additional shipping may apply.
Caution: This bed is refurbished and is intended for educational and training use only.