Quantum Levitation Maglev Kit
Supplier: QUANTUM EXPERIENCE LTD
307
470306-204EA 7501.59 USD
470306-204
Quantum Levitation Maglev Kit
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The maglev kit is perfect for large audience demonstrations at science museums, science fairs and student classes.
  • Easy to use
  • Highly durable
The circular frictionless motion never seizes to amaze.
When used with the new resistance thermometer equipped Ed levitator, the handheld magnetic device serves as a unique experimentation platform where quantum phenomena such as the Meissner effect and flux pinning can be investigated.
Delivery information: The kit is supplied with large and medium Quantum Levitators which can be levitated simultaneously in a double levitation configuration. Both suspension below the track and levitation above it can be demonstrated. A handheld magnetic device (Starter Kit) is also included for a complete experience.