Go Direct™ Elementary Packages
Supplier: Vernier Software & Technology
GDP-EL-DX
470305-410EA 731.99 USD
470305-410 470305-412
Go Direct™ Elementary Packages
Data Loggers
We offer different packages to meet your needs and budget for the elementary classroom.
  • Starter Package includes temperature, light and color, motion detector probes
  • Elementary Science with Vernier contains 43 fun and engaging experiments
  • Deluxe Package includes (in addition to starter package) three-axis magnetic field, gas pressure, voltage, force and acceleration probes
The starter package is great way to get started, and the deluxe package includes all the sensors needed for the experiments in the Elementary Science with Vernier item 470017-666.
Our Go Direct™ Probes connects directly to your mobile device, Chromebook, or computer using our free Graphical Analysis™ 4 app-no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via bluetooth, allowing you to choose the solution best for your classroom or laboratory.
Delivery information: Includes Go Direct® temperature probe, Go Direct® light and color sensor, Go Direct® motion detector, Go Direct® three-Axis magnetic field sensor, Go Direct® gas pressure sensor, gas pressure sensor bulk, Go Direct® voltage probe, Go Direct® force and acceleration sensor