Solar Balloon
Supplier: Tedco
80010
470305-374CS 134.25 USD
470305-374
Solar Balloon
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Great Intro For Solar Energy!
  • Teach Bernoulli's and Pascal's principles
  • Includes science learning guide
  • Gigantic size allows everyone to participate!
Giant-sized solar science fun! This 50 foot long balloon soars high in the sky powered by the sun. Demonstrates buoyancy, convection, thermodynamics and solar power. Teaches Bernoulli's Principle and Pascals Principle. Great for science classes and the perfect start to a science project. Bright direct sunlight on a calm, cool day has the Solar Balloon soaring in just minutes. Includes one 50 foot long by 29" diameter Solar Balloon, 400 feet of string, complete instructions and science learning guide. Made in USA. For ages 14+.