WHEATON® Dispensing Bottle, LDPE, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
211194
470304-924CS 498.05 USD
470304-924
WHEATON® Dispensing Bottle, LDPE, DWK Life Sciences
Bottles Plastic Bottles
LDPE dispensing bottle. Raise the spout to dispense contents and depress spout to seal the bottle.
  • Made from low density polyethylene
  • Translucent color
  • Very flexible with high impact strength
  • Excellent for mild and strong buffers, good chemical resistance