Universal Indicator Color Charts
Supplier: Ward's Science
XU0011-C XU0012-C XU0013-C
470304-892EA 12.55 USD
470304-892 470304-894 470304-896
Universal Indicator Color Charts
Educational Materials Chemistry Educational Materials Acids and Bases Learning Activities
Easily determine pH with the universal indicator chart.
  • Three sizes available
  • Complete color/pH scale
The universal indicator color chart is used to determine the pH of a solution. The charts display the colors and pH values of each pH unit from ≤3 to ≥10. The charts are available as an overhead transparency, 8.5×11" sheet and smaller chart cards.