Ultraviolet Light
Supplier: United Scientific Supplies
AISUVLT1
470304-018EA 15.4 USD
470304-018
Ultraviolet Light
Lights UV Lights, Lamps and Bulbs
High Quality for the Classroom.
  • Built-in LED flashlight
This high-quality 6", 4 W handheld blacklight is ideal for forensics or illuminating fluorescent and blacklight reactive items. Requires 4 AA batteries (not included).