Tissue Floating Bath for Slide-making
Supplier: C & A Scientific
XH-1001
470303-892EA 301.45 USD
470303-892
Tissue Floating Bath for Slide-making
Baths Tissue Baths
The low profile, 2.3 l, black, finished interior chamber of this unit reduces glare, improving visibility of the tissue specimens.
  • Black interior chamber reduces glare
  • Glazed glass lid
  • Durable, fiberglass insulated body prevents heat loss
  • Detachable power cord for easy cleaning
It includes a glazed glass lid for easy viewing of interior, and the top ledge allows space for preheating or for evaporating away any remaining water on the glass slides. Its durable, fiberglass insulated body prevents heat loss and fluctuations in temperature for uniform surface heating. Includes non-stick coated chamber and detachable power cord for easy cleaning.
Interior size: 8.25×2.5"