Go Direct™ Surface Temperature
Supplier: Vernier Software & Technology
GDX-ST
470303-846EA 98.99 USD
470303-846
Go Direct™ Surface Temperature
Data Loggers
Designed for use in situations in which low thermal mass or flexibility is required, the Go Direct Surface Temperature Sensor connects wirelessly via Bluetooth® or wired via USB to your device.
  • Investigate the effect of temperature on solar panel output
  • Explore passive solar heating or solar collectors
  • Examine the warming function of nasal passageways
  • Discover the effect of vascularity on skin temperature recovery
This sensor has an exposed thermistor that results in an extremely rapid response time, and this design allows for use in air and water.
The Go Direct robust rechargeable battery offers long battery life and provides always-ready operation. Monitor battery life directly from our free Graphical Analysis™ 4 app. Should the battery run low, simply connect the sensor to the charging cable and keep on collecting-no interruptions or inconvenience to you or your students.
Our Go Direct sensors connect directly to your mobile device, Chromebook™, or computer using our free Graphical Analysis™ 4 app-no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth wireless technology, allowing you to choose the solution best for your classroom or laboratory.
Ordering information: Important–For use in air and water only. For temperature measurements in harsher environments that require a more durable probe, we recommend the Go Direct® Temperature Probe.
Delivery information: Includes Go Direct surface temperature sensor, micro USB cable, quick-start guide.