Wax Trimmer
Supplier: C & A Scientific
XH-90D
470303-816EA 630.65 USD
470303-816
Wax Trimmer
Paraffin Wax Trimmers and Dispensers
This wax trimmer is compact but powerful.
  • Disposable tray liner allows easy disposal or reuse of wax runoff
  • Digital display allows for user to adjust temperature to exact values
  • Can trim multiple cassettes simultaneously
  • Red LED light alerts users when unit is on and heating
The magnetically secured tray and included disposable tray liner collects melted paraffin, allowing for easy and clean disposal. This efficient and practical appliance is best used for removig excess paraffin from tissue cassettes.
Overall size of the unit is 11.25×5×8" including the tray. Surface size is 9×5". Requires 150 W power.
Delivery information: 5 disposable tray liners included. Additional liners sold separately.