Medline® Pocket Otoscope
Supplier: MEDLINE INDUSTRIES, INC.
MDS2111203
470303-258EA 425 USD
470303-258
Medline® Pocket Otoscope
Opthalmoscopes
Small and powerful.
  • Compact AA size
  • Consistent light output
  • Brilliant LED light source
This pocket otoscope has all the features you would expect on larger models, with the convenience of an AA sized handle. Unlike some pocket-sized scopes, this otoscope has a metal housing and a glass viewing window. The 3.7 V LED provides consistent illumination throughout the 50-hour battery life. The small size fits perfectly in a pocket, or a field bag. If you want some extra travel protection it even comes with a hard-shell case to prevent damage from harder shocks.
Ordering information: An insufflation bulb is not included. If you would like to purchase one, please contact customer service. Does not include batteries.