Medline® Otoscope/Ophthalmoscope Desk Set
Supplier: MEDLINE INDUSTRIES, INC.
MDS107031
470303-256EA 1932.15 USD
470303-256
Medline® Otoscope/Ophthalmoscope Desk Set
Opthalmoscopes
Designed for demanding everyday use.
  • LED illumination for otoscopes
  • 3.5 V xenon illumination for ophthalmological instruments supply daylight conditions
  • With desktop charger
Always have your instruments handy with this convenient desktop Otoscope and Ophthalmoscope bundle. The slim AA sized handles are easy to grip and fit right in your pocket out of the way. When they aren't being used they store easily in the compact charging station, so they are always ready.
The ophthalmoscope uses a diopter wheel with 29 corrective lenses, an easy to operate aperture wheel and a xenon bulb for the best possible illumination. The Otoscope features a 3X magnification swiveling optical glass, and a Fiberoptic LED light source.