Zinc Carbonate
Supplier: ALDON CORP SE
ZZ0050-500G
470303-072EA
63.65
USD
470303-072
Zinc Carbonate
Chemicals for Science Education
CAS Number: 5970-47-8
Formula Weight: 548.96
Formula: [ZnCO3]2 [Zn(OH2]3
Density (g/mL): 3.5
Solubility: Slightly soluble in water
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...