Wright's Stain Buffer
Supplier: ALDON CORP SE
CAS Number: Mixture
Formula Weight: Mixture
Formula: Mixture
Shelf Life (months): 36
Storage: Green
Caution: Please Note: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...